(Z)-5-(4-(Dimethylamino)benzylidene)-2-thioxothiazolidin-4-one 80%+

Supplier: Ambeed
Warning

A960289-25G A960289-1G A960289-5G
76656-154EA 42 USD
76656-154 77623-192 77623-194
(Z)-5-(4-(Dimethylamino)benzylidene)-2-thioxothiazolidin-4-one 80%+
(Z)-5-(4-(Dimethylamino)benzylidene)-2-thioxothiazolidin-4-one

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂OS₂
MDL Number: MFCD00005274
CAS Number: 1134203-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR