tert-Butyl 8-bromooctanoate 97%

Supplier: Ambeed
Warning

A956836-1G A956836-5G A956836-250MG
76656-126EA 154.9 USD
76656-126 76656-128 77599-302
tert-Butyl 8-bromooctanoate 97%
tert-Butyl 8-bromooctanoate

Precaución: for Research Use Only

Formula: C₁₂H₂₃BrO₂
MW: 279.21 g/mol
MDL Number: MFCD27928608
CAS Number: 77383-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR