5-(N,N-Dimethylsulfamoyl)-N-(5-methylthiazol-2-yl)-2-(pyrrolidin-1-yl)benzamide 99%

Supplier: Ambeed
Warning

A956586-10MG A956586-1MG A956586-25MG A956586-5MG A956586-50MG A956586-100MG
76656-118EA 39 USD
76656-118 76656-120 76656-122 76656-124 77623-184 77784-412
5-(N,N-Dimethylsulfamoyl)-N-(5-methylthiazol-2-yl)-2-(pyrrolidin-1-yl)benzamide 99%
5-(N,N-Dimethylsulfamoyl)-N-(5-methylthiazol-2-yl)-2-(pyrrolidin-1-yl)benzamide

Precaución: for Research Use Only

Formula: C₁₇H₂₂N₄O₃S₂
MDL Number: MFCD06358908
CAS Number: 790702-57-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR