1-Ethynyl-4-(phenylethynyl)benzene 98% (by GC)

Supplier: Ambeed
Warning

Synonyms: 1-(4-Ethynylphenyl)-2-phenylacetylene, 4-Ethynyldiphenylacetylene

A955699-100MG A955699-250MG A955699-1G
76656-114EA 37.14 USD
76656-114 76656-116 77595-542
1-Ethynyl-4-(phenylethynyl)benzene 98% (by GC)
1-Ethynyl-4-(phenylethynyl)benzene

Precaución: for Research Use Only

Formula: C₁₆H₁₀
MW: 202.26 g/mol
MDL Number: MFCD27665710
CAS Number: 92866-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR