6'-Amino-4'-ethyl-5'-(4-hydroxyphenyl)-N-methyl-[3,3'-bipyridine]-6-carboxamide 98%

Supplier: Ambeed
Warning

Synonyms: GNE-6776

A955293-1MG A955293-25MG A955293-5MG A955293-100MG A955293-250MG A955293-50MG
76656-102EA 27.59 USD
76656-102 76656-104 76656-106 77623-178 77623-180 77623-182
6'-Amino-4'-ethyl-5'-(4-hydroxyphenyl)-N-methyl-[3,3'-bipyridine]-6-carboxamide 98%
6'-Amino-4'-ethyl-5'-(4-hydroxyphenyl)-N-methyl-[3,3'-bipyridine]-6-carboxamide

Precaución: for Research Use Only

Formula: C₂₀H₂₀N₄O₂
MW: 348.4 g/mol
MDL Number: MFCD31657364
CAS Number: 2009273-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR