1-(2-Methylquinolin-6-yl)ethanone 95+%

Supplier: Ambeed
Warning

A955280-1G A955280-250MG
76656-098EA 203.7 USD
76656-098 76656-100
1-(2-Methylquinolin-6-yl)ethanone 95+%
1-(2-Methylquinolin-6-yl)ethanone

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO
MW: 185.22 g/mol
MDL Number: MFCD11553498
CAS Number: 116632-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR