Methyl 4-fluoro-2-hydroxy-3-nitrobenzoate 97%

Supplier: Ambeed
Danger

A955271-100MG A955271-1G A955271-250MG
76656-092EA 91 USD
76656-092 76656-094 76656-096
Methyl 4-fluoro-2-hydroxy-3-nitrobenzoate 97%
Methyl 4-fluoro-2-hydroxy-3-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₆FNO₅
MDL Number: MFCD18074446
CAS Number: 1263283-00-6
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR