N,5-Dimethylpyridin-2-amine 95%

Supplier: Ambeed
Warning

A954863-250MG A954863-5G
76656-078EA 45.62 USD
76656-078 76656-080
N,5-Dimethylpyridin-2-amine 95%
N,5-Dimethylpyridin-2-amine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂
MW: 122.168 g/mol
MDL Number: MFCD06411230
CAS Number: 45715-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR