2,8-Difluoro-5-(trifluoromethyl)-5H-dibenzo[b,d]thiophen-5-ium trifluoromethanesulfonate 98% (by HPLC)

Supplier: Ambeed
Warning

Synonyms: 2,8-Difluoro-5-(trifluoromethyl)-5H-dibenzo[b,d]thiophen-5-ium triflate, Umemoto's Reagent II

A953556-1G A953556-25G A953556-5G
76656-060EA 36.08 USD
76656-060 76656-062 76656-064
2,8-Difluoro-5-(trifluoromethyl)-5H-dibenzo[b,d]thiophen-5-ium trifluoromethanesulfonate 98% (by HPLC)
2,8-Difluoro-5-(trifluoromethyl)-5H-dibenzo[b,d]thiophen-5-ium trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₁₄H₆F₈O₃S₂
MW: 438.31 g/mol
MDL Number: MFCD30725753
CAS Number: 1961266-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR