4-(4-(Trifluoromethoxy)phenoxy)piperidine hydrochloride 95%

Supplier: Ambeed
Warning

A952959-1G A952959-250MG A952959-5G
76656-054EA 45.62 USD
76656-054 76656-056 76656-058
4-(4-(Trifluoromethoxy)phenoxy)piperidine hydrochloride 95%
4-(4-(Trifluoromethoxy)phenoxy)piperidine hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₅ClF₃NO₂
MW: 297.7000 g/mol
MDL Number: MFCD30721028
CAS Number: 1501963-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR