Edonerpic maleate 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
T-817, 1-(3-(2-(benzo[b]thiophen-5-yl)ethoxy)propyl)azetidin-3-ol maleate
A952655-100MG
A952655-10MG
A952655-1MG
A952655-25MG
A952655-50MG
A952655-5MG
76656-042EA
1935
USD
76656-042
76656-044
76656-046
76656-048
76656-050
76656-052
Edonerpic maleate 99%
Edonerpic maleate
Precaución: for Research Use Only
Formula:
C₂₀H₂₅F₅NO₆S MW: 407.48 g/mol |
CAS Number:
519187-97-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...