Lithium (R)-1-(tert-butoxycarbonyl)aziridine-2-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A951032-1G
A951032-250MG
76656-026EA
941.02
USD
76656-026
76656-028
Lithium (R)-1-(tert-butoxycarbonyl)aziridine-2-carboxylate 97%
Lithium (R)-1-(tert-butoxycarbonyl)aziridine-2-carboxylate
Precaución: for Research Use Only
Formula:
C₈H₁₂LiNO₄ MW: 193.1262 g/mol |
MDL Number:
MFCD31620800 CAS Number: 2173637-01-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...