Ethyl 2-(benzo[c][1,2,5]thiadiazol-5-yl)acetate 97%

Supplier: Ambeed
Warning

A950552-1G A950552-250MG A950552-5G
76656-012EA 49.87 USD
76656-012 76656-014 76656-016
Ethyl 2-(benzo[c][1,2,5]thiadiazol-5-yl)acetate 97%
Ethyl 2-(benzo[c][1,2,5]thiadiazol-5-yl)acetate

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂S
MW: 222.2600 g/mol
MDL Number: MFCD28127334
CAS Number: 195505-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR