2-Amino-3-chloro-5-methylphenol 97%

Supplier: Ambeed
Warning

A950018-1G A950018-5G
76656-000EA 135 USD
76656-000 76656-002
2-Amino-3-chloro-5-methylphenol 97%
2-Amino-3-chloro-5-methylphenol

Precaución: for Research Use Only

Formula: C₇H₈ClNO
MDL Number: MFCD28551485
CAS Number: 1780449-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR