3,6,9,12-Tetraoxatetradecane-1,14-dioic Acid 95%

Supplier: Ambeed
Danger

Synonyms: CH2COOH-PEG3-CH2COOH

A949151-1G A949151-250MG A949151-5G A949151-100MG
76655-978EA 159.14 USD
76655-978 76655-980 76655-982 77623-176
3,6,9,12-Tetraoxatetradecane-1,14-dioic Acid 95%
3,6,9,12-Tetraoxatetradecane-1,14-dioic Acid

Precaución: for Research Use Only

Formula: C₁₀H₁₈O₈
MW: 266.2451 g/mol
MDL Number: MFCD21363215
CAS Number: 32775-08-9
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR