Ethyl 2-(4-((4-nitrobenzyl)oxy)benzyl)thiazolidine-4-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: SN 6

A948730-10MG A948730-1MG A948730-25MG A948730-5MG
76655-956EA 50.93 USD
76655-956 76655-958 76655-960 76655-962
Ethyl 2-(4-((4-nitrobenzyl)oxy)benzyl)thiazolidine-4-carboxylate 98%
Ethyl 2-(4-((4-nitrobenzyl)oxy)benzyl)thiazolidine-4-carboxylate

Precaución: for Research Use Only

Formula: C₂₀H₂₂N₂O₅S
MW: 402.4641 g/mol
MDL Number: MFCD06798326
CAS Number: 415697-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR