(Isocyanoimino)triphenylphosphorane 98%

Supplier: Ambeed
Warning

A944419-1G A944419-5G A944419-10G
76655-924EA 11.67 USD
76655-924 76655-926 77573-880
(Isocyanoimino)triphenylphosphorane 98%
(Isocyanoimino)triphenylphosphorane

Precaución: for Research Use Only

Formula: C₁₉H₁₅N₂P
MW: 302.32 g/mol
MDL Number: MFCD09038528
CAS Number: 73789-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR