3-(2-(6-(tert-Butyl)-1H-benzo[d]imidazol-2-yl)ethyl)cyclobutanone hydrochloride 97%

Supplier: Ambeed
Warning

A943892-100MG A943892-250MG
76655-904EA 95 USD
76655-904 76655-906
3-(2-(6-(tert-Butyl)-1H-benzo[d]imidazol-2-yl)ethyl)cyclobutanone hydrochloride 97%
3-(2-(6-(tert-Butyl)-1H-benzo[d]imidazol-2-yl)ethyl)cyclobutanone hydrochloride

Precaución: for Research Use Only

Formula: C₁₇H₂₃ClN₂O
MW: 306.8300 g/mol
CAS Number: 1628338-77-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR