(S)-Methyl 3-(3-bromophenyl)-2-((tert-butoxycarbonyl)amino)propanoate 98%

Supplier: Ambeed
Warning

A943881-100MG A943881-1G A943881-250MG
76655-898EA 106.09 USD
76655-898 76655-900 76655-902
(S)-Methyl 3-(3-bromophenyl)-2-((tert-butoxycarbonyl)amino)propanoate 98%
(S)-Methyl 3-(3-bromophenyl)-2-((tert-butoxycarbonyl)amino)propanoate

Precaución: for Research Use Only

Formula: C₁₅H₂₀BrNO₄
MW: 358.23 g/mol
MDL Number: MFCD22418905
CAS Number: 546115-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR