4'-(Bromomethyl)-2-chlorobiphenyl 90%

Supplier: Ambeed
Warning

Synonyms: 4-(2-Chlorophenyl)benzyl bromide, 4'-(Bromomethyl)-2-chloro-1,1'-biphenyl

A943594-100MG A943594-1G A943594-250MG
76655-880EA 98.67 USD
76655-880 76655-882 76655-884
4'-(Bromomethyl)-2-chlorobiphenyl 90%
4'-(Bromomethyl)-2-chlorobiphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀BrCl
MW: 281.576 g/mol
MDL Number: MFCD15146809
CAS Number: 210113-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR