IDH-305 98%

Supplier: Ambeed
Warning

Synonyms: (R)-4-((S)-1-Fluoroethyl)-3-(2-(((S)-1-(4-methyl-2'-(trifluoromethyl)-[3,4'-bipyridin]-6-yl)ethyl)amino)pyrimidin-4-yl)oxazolidin-2-one

A943584-10MG A943584-1MG A943584-5MG
76655-874EA 289.63 USD
76655-874 76655-876 76655-878
IDH-305 98%
IDH-305

Precaución: for Research Use Only

Formula: C₂₃H₂₂F₄N₆O₂
MW: 490.45 g/mol
MDL Number: MFCD31630851
CAS Number: 1628805-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR