2-Amino-5-tert-butyl-benzonitrile 95%

Supplier: Ambeed
Warning

A943255-1G A943255-5G A943255-25G
76655-856EA 26 USD
76655-856 76655-858 77784-391
2-Amino-5-tert-butyl-benzonitrile 95%
2-Amino-5-tert-butyl-benzonitrile

Precaución: for Research Use Only

Formula: C₁₁H₁₄N₂
MDL Number: MFCD06408810
CAS Number: 874814-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR