3'-Fluoro-4'-nitroacetanilide 98%

Supplier: Ambeed
Danger

Synonyms: N-(3-fluoro-4-nitrophenyl)acetamide

A943212-1G A943212-250MG A943212-5G
76655-850EA 53.05 USD
76655-850 76655-852 76655-854
3'-Fluoro-4'-nitroacetanilide 98%
3'-Fluoro-4'-nitroacetanilide

Precaución: for Research Use Only

Formula: C₈H₇FN₂O₃
MW: 198.1512 g/mol
MDL Number: MFCD01463345
CAS Number: 345-30-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR