7-Amino-8-bromo-3,4-dihydronaphthalen-1(2H)-one 97%

Supplier: Ambeed
Warning

A1350478-250MG A1350478-5G A1350478-1G
76658-176EA 55 USD
76658-176 77549-626 77592-552
7-Amino-8-bromo-3,4-dihydronaphthalen-1(2H)-one 97%
7-Amino-8-bromo-3,4-dihydronaphthalen-1(2H)-one

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrNO
CAS Number: 214697-98-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR