2-(Dicyclohexylphosphino)-N,N-dimethyl[1,1-biphenyl]-4-amine 98%

Supplier: Ambeed
Warning

A943093-100MG A943093-1G A943093-250MG A943093-5G
76655-836EA 26 USD
76655-836 76655-838 76655-840 77618-604
2-(Dicyclohexylphosphino)-N,N-dimethyl[1,1-biphenyl]-4-amine 98%
2-(Dicyclohexylphosphino)-N,N-dimethyl[1,1-biphenyl]-4-amine

Precaución: for Research Use Only

Formula: C₂₆H₃₆NP
MDL Number: MFCD28400094
CAS Number: 1185899-00-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR