2-(4-((2-Methoxyethoxy)methyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%

Supplier: Ambeed
Warning

A942880-1G A942880-250MG A942880-5G
76655-818EA 123.07 USD
76655-818 76655-820 76655-822
2-(4-((2-Methoxyethoxy)methyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(4-((2-Methoxyethoxy)methyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₆H₂₅BO₄
MW: 292.178 g/mol
MDL Number: MFCD20527108
CAS Number: 1580442-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR