Propyl methanesulfonate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methanesulphonic acid Propyl Ester
A942867-100G
A942867-25G
A942867-5G
76655-812EA
332.07
USD
76655-812
76655-814
76655-816
Propyl methanesulfonate 98%
Propyl methanesulfonate
Precaución: for Research Use Only
Formula:
C₄H₁₀O₃S MW: 138.18 g/mol |
MDL Number:
MFCD00007561 CAS Number: 1912-31-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...