(4-Bromo-2,6-difluorophenyl)trimethylsilane 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2,6-difluoro-1-(trimethylsilyl)benzene

A940624-100MG A940624-1G A940624-250MG A940624-5G
76655-776EA 74 USD
76655-776 76655-778 76655-780 76655-782
(4-Bromo-2,6-difluorophenyl)trimethylsilane 98%
(4-Bromo-2,6-difluorophenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₉H₁₁BrF₂Si
MDL Number: MFCD18642935
CAS Number: 184910-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR