Tris(dibenzylideneacetone)dipalladium(0) 98%

Supplier: Ambeed
Danger

Synonyms: Pd2(dba)3, Pd2dba3, Tris(dibenzylideneacetonyl)bis-palladium, Bis[tris(dibenzylideneacetone)palladium(0)]

A939841-10G A939841-1G A939841-250MG A939841-25G A939841-5G A939841-100G
76655-748EA 477.41 USD
76655-748 76655-750 76655-752 76655-754 76655-756 77749-766
Tris(dibenzylideneacetone)dipalladium(0) 98%
Tris(dibenzylideneacetone)dipalladium(0)

Precaución: for Research Use Only

Formula: C₅₁H₄₂O₃Pd₂
MW: 915.72 g/mol
MDL Number: MFCD00013310
CAS Number: 51364-51-3
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR