But-2-ene-1,4-diamine, N-BOC protected 95%

Supplier: Ambeed
Warning

A939469-1G A939469-25G A939469-100G A939469-5G A939469-250MG
76655-738EA 17 USD
76655-738 76655-740 77552-738 77552-740 77599-280
But-2-ene-1,4-diamine, N-BOC protected 95%
But-2-ene-1,4-diamine, N-BOC protected

Precaución: for Research Use Only

Formula: C₉H₁₈N₂O₂
MDL Number: MFCD09752966
CAS Number: 146394-99-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR