5-Bromo-1,3-benzenedicarbonyl dichloride 98%

Supplier: Ambeed
Danger

A938662-1G A938662-250MG A938662-25G A938662-5G
76655-726EA 35 USD
76655-726 76655-728 76655-730 76655-732
5-Bromo-1,3-benzenedicarbonyl dichloride 98%
5-Bromo-1,3-benzenedicarbonyl dichloride

Precaución: for Research Use Only

Formula: C₈H₃BrCl₂O₂
MDL Number: MFCD28009845
CAS Number: 57863-69-1
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR