6-Amino-3-bromo-2-fluorobenzoic acid 97%

Supplier: Ambeed
Warning

A938481-5G A938481-1G A938481-25G A938481-100G
76655-724EA 48 USD
76655-724 77749-762 77749-763 77749-761
6-Amino-3-bromo-2-fluorobenzoic acid 97%
6-Amino-3-bromo-2-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅BrFNO₂
MDL Number: MFCD20482512
CAS Number: 1036756-03-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR