1,3-Bis[4-(trifluoromethyl)phenyl]thiourea 97% (by HPLC, total nitrogen)

Supplier: Ambeed
Warning

A938347-10G A938347-25G A938347-5G A938347-1G
76655-718EA 232 USD
76655-718 76655-720 76655-722 77749-760
1,3-Bis[4-(trifluoromethyl)phenyl]thiourea 97% (by HPLC, total nitrogen)
1,3-Bis[4-(trifluoromethyl)phenyl]thiourea

Precaución: for Research Use Only

Formula: C₁₅H₁₀F₆N₂S
MDL Number: MFCD18191886
CAS Number: 1744-07-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR