[3,5-Bis(4-fluorobenzoyl)phenyl](4-fluorophenyl)methanone 90%

Supplier: Ambeed
Warning

A937551-100MG A937551-1G A937551-250MG A937551-5G
76655-710EA 50.93 USD
76655-710 76655-712 76655-714 76655-716
[3,5-Bis(4-fluorobenzoyl)phenyl](4-fluorophenyl)methanone 90%
[3,5-Bis(4-fluorobenzoyl)phenyl](4-fluorophenyl)methanone

Precaución: for Research Use Only

Formula: C₂₇H₁₅F₃O₃
MW: 444.4014 g/mol
MDL Number: MFCD00683306
CAS Number: 267668-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR