SU 3327 98% (by HPLC)

Supplier: Ambeed
Warning

Synonyms: 2-Amino-5-[(5-nitro-2-thiazolyl)thio]-1,3,4-thiadiazole, SU 3327, 5-((5-Nitrothiazol-2-yl)thio)-1,3,4-thiadiazol-2-amine, SU3327

A937290-250MG A937290-50MG A937290-100MG
76655-700EA 192.03 USD
76655-700 77599-276 77623-170
SU 3327 98% (by HPLC)
SU 3327

Precaución: for Research Use Only

Formula: C₅H₃N₅O₂S₃
MW: 261.30 g/mol
MDL Number: MFCD01927019
CAS Number: 40045-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR