Spiro[3.5]nonan-6-one 98%

Supplier: Ambeed
Warning

A936965-100MG A936965-250MG A936965-50MG
76655-694EA 63 USD
76655-694 76655-696 76655-698
Spiro[3.5]nonan-6-one 98%
Spiro[3.5]nonan-6-one

Precaución: for Research Use Only

Formula: C₉H₁₄O
MDL Number: MFCD24705291
CAS Number: 74064-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR