N-(4-Chloro-2-isopropoxybenzyl)-5-phenyl-[1,2,4]triazolo[1,5-a]pyrimidin-7-amine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
AF64394
A935740-10MG
A935740-1MG
A935740-25MG
A935740-50MG
A935740-5MG
76655-676EA
210.06
USD
76655-676
76655-678
76655-680
76655-682
76655-684
N-(4-Chloro-2-isopropoxybenzyl)-5-phenyl-[1,2,4]triazolo[1,5-a]pyrimidin-7-amine 98%
N-(4-Chloro-2-isopropoxybenzyl)-5-phenyl-[1,2,4]triazolo[1,5-a]pyrimidin-7-amine
Precaución: for Research Use Only
Formula:
C₂₁H₂₀ClN₅O MW: 393.8694 g/mol |
MDL Number:
MFCD28411712 CAS Number: 1637300-25-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...