1-Butyl-3-methylimidazoliumtrifluoroacetate 97% (by titrimetric analysis)

Supplier: Ambeed
Warning

Synonyms: 1-Butyl-3-methylimidazolium Trifluoroacetate

A935110-1G A935110-25G A935110-5G
76655-664EA 5.31 USD
76655-664 76655-666 76655-668
1-Butyl-3-methylimidazoliumtrifluoroacetate 97% (by titrimetric analysis)
1-Butyl-3-methylimidazoliumtrifluoroacetate

Precaución: for Research Use Only

Formula: C₁₀H₁₅F₃N₂O₂
MW: 252.24 g/mol
MDL Number: MFCD08458921
CAS Number: 174899-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR