7-Bromo-6-fluoro-1H-indazole 97%

Supplier: Ambeed
Warning

A934711-100MG A934711-250MG A934711-5G A934711-1G
76655-652EA 55 USD
76655-652 76655-654 76655-656 77573-868
7-Bromo-6-fluoro-1H-indazole 97%
7-Bromo-6-fluoro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrFN₂
MDL Number: MFCD23705437
CAS Number: 1427377-75-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR