3,4-Dichloro-5-fluoroaniline 97%

Supplier: Ambeed
Danger

A934144-250MG A934144-1G A934144-100MG A934144-5G
76655-650EA 18 USD
76655-650 77595-510 77623-168 77784-381
3,4-Dichloro-5-fluoroaniline 97%
3,4-Dichloro-5-fluoroaniline

Precaución: for Research Use Only

Formula: C₆H₄Cl₂FN
MDL Number: MFCD22376609
CAS Number: 35754-38-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR