2,4-Dichloro-N-(4-(phenylcarbamoyl)phenyl)benzamide 98%

Supplier: Ambeed
Warning

Synonyms: CID 1375606

A934025-100MG A934025-10MG A934025-1MG A934025-25MG A934025-50MG A934025-5MG
76655-638EA 140.04 USD
76655-638 76655-640 76655-642 76655-644 76655-646 76655-648
2,4-Dichloro-N-(4-(phenylcarbamoyl)phenyl)benzamide 98%
2,4-Dichloro-N-(4-(phenylcarbamoyl)phenyl)benzamide

Precaución: for Research Use Only

Formula: C₂₀H₁₄Cl₂N₂O₂
MW: 385.24 g/mol
MDL Number: MFCD00369571
CAS Number: 313493-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR