SB-297006 97%

Supplier: Ambeed
Danger

A933645-10MG A933645-1MG A933645-25MG A933645-50MG A933645-5MG
76655-626EA 57 USD
76655-626 76655-628 76655-630 76655-632 76655-634
SB-297006 97%
SB-297006

Precaución: for Research Use Only

Formula: C₁₈H₁₈N₂O₅
MDL Number: MFCD19690954
CAS Number: 58816-69-6
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR