5-(4-Methylpiperazin-1-yl)-2-nitrobenzaldehyde 95%

Supplier: Ambeed
Warning

A933412-100MG A933412-1G A933412-250MG A933412-5G
76655-618EA 6 USD
76655-618 76655-620 76655-622 76655-624
5-(4-Methylpiperazin-1-yl)-2-nitrobenzaldehyde 95%
5-(4-Methylpiperazin-1-yl)-2-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₁₅N₃O₃
MDL Number: MFCD20485290
CAS Number: 904895-82-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR