3,4-Difluoro-5-methylaniline 97%

Supplier: Ambeed
Warning

A933361-1G A933361-250MG
76655-614EA 426.49 USD
76655-614 76655-616
3,4-Difluoro-5-methylaniline 97%
3,4-Difluoro-5-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇F₂N
MW: 143.134 g/mol
MDL Number: MFCD22490569
CAS Number: 1505944-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR