2-{bicyclo[1.1.1]pentan-1-yl}-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%
Supplier: Ambeed
|
Warning
|
A933043-100MG
A933043-1G
A933043-250MG
A933043-50MG
76655-606EA
317.21
USD
76655-606
76655-608
76655-610
76655-612
2-{bicyclo[1.1.1]pentan-1-yl}-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%
2-{bicyclo[1.1.1]pentan-1-yl}-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only
Formula:
C₁₁H₁₉BO₂ MW: 194.08 g/mol |
MDL Number:
MFCD31705662 CAS Number: 2152645-07-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...