Ethyl 4-cyano-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 95%

Supplier: Ambeed
Warning

A932902-1G A932902-250MG
76655-602EA 189 USD
76655-602 76655-604
Ethyl 4-cyano-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 95%
Ethyl 4-cyano-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

Precaución: for Research Use Only

Formula: C₁₆H₂₀BNO₄
MDL Number: MFCD22494140
CAS Number: 1126479-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR