Methyl 2-(3,4,4-trifluorobut-3-ene-1-sulfonyl)acetate 90%

Supplier: Ambeed
Warning

A931965-100MG A931965-250MG
76655-572EA 138.98 USD
76655-572 76655-574
Methyl 2-(3,4,4-trifluorobut-3-ene-1-sulfonyl)acetate 90%
Methyl 2-(3,4,4-trifluorobut-3-ene-1-sulfonyl)acetate

Precaución: for Research Use Only

Formula: C₇H₉F₃O₄S
MW: 246.2042 g/mol
MDL Number: MFCD01443882
CAS Number: 339020-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR