6-(Methylsulfonyl)picolinaldehyde 95%

Supplier: Ambeed
Warning

A929561-100MG A929561-250MG A929561-50MG
76655-564EA 106.09 USD
76655-564 76655-566 76655-568
6-(Methylsulfonyl)picolinaldehyde 95%
6-(Methylsulfonyl)picolinaldehyde

Precaución: for Research Use Only

Formula: C₇H₇NO₃S
MW: 185.2000 g/mol
MDL Number: MFCD30188729
CAS Number: 1780013-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR