3-Methyldihydro-2H-thiopyran-4(3H)-one 95%

Supplier: Ambeed
Warning

A928396-100MG A928396-250MG
76655-542EA 116.7 USD
76655-542 76655-544
3-Methyldihydro-2H-thiopyran-4(3H)-one 95%
3-Methyldihydro-2H-thiopyran-4(3H)-one

Precaución: for Research Use Only

Formula: C₆H₁₀OS
MW: 130.21 g/mol
MDL Number: MFCD20483860
CAS Number: 38486-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR