2-(3-((3-Chlorophenoxy)methyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 96%

Supplier: Ambeed
Warning

A927413-100MG A927413-1G A927413-250MG
76655-522EA 36.08 USD
76655-522 76655-524 76655-526
2-(3-((3-Chlorophenoxy)methyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 96%
2-(3-((3-Chlorophenoxy)methyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₉H₂₂BClO₃
MW: 344.6402 g/mol
MDL Number: MFCD20491435
CAS Number: 2096332-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR